Gene Transcripts

Gene Symbol
POM121C
Entrez Gene ID
100101267
Gene Name
POM121 transmembrane nucleoporin C
Gene Aliases
POM121-2
Chromosome Location
Chr. 7: 75416781 - 75486288 on Build GRCh38
UniGene ID
Hs.712563
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099415.2 NP_001092885.2 13 3148
GenBank mRNA AK293305.1 - 1 691
AK293781.1 - 9 2204
AK311407.1 - 1 42
BC082993.1 - 13 3125

View Full Product Details