Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099415.2 | NP_001092885.2 | 14, 15 | 3750 | |
GenBank mRNA | AK127026.1 | - | 1, 2 | 1968 | |
AK293305.1 | - | 2, 3 | 1293 | ||
AK293781.1 | - | 10, 11 | 2806 | ||
AK311407.1 | - | 2, 3 | 644 | ||
BC082993.1 | - | 14, 15 | 3727 | ||
BU189720.1 | - | 2, 3 | 401 | ||
BX649044.1 | - | 1, 2 | 324 |