Gene Transcripts

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
Chromosome Location
Chr. 19: 19645198 - 19663694 on Build GRCh38
UniGene ID
Hs.501794
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020410.2 NP_065143.2 23 3152
GenBank mRNA AB058728.1 - 23 3122
AF288687.1 - 23 2770
AK026044.1 - 7 1302
AK056420.1 - 23 3128
AK074159.1 - 14 3937
AK074798.1 - 11 1657
AK074920.1 - 12 1732
AK095287.1 - 6 2325
AK123780.1 - 6 1496
AK125861.1 - 7 3449
AK130125.1 - 14 1792
AK172778.1 - 22 2583
AL834180.1 - 1 1382
BC009302.2 - 5 596
BC069211.1 - 4 486

View Full Product Details