Gene Transcripts

Gene Symbol
HNRNPA1
Entrez Gene ID
3178
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Gene Aliases
ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1
Chromosome Location
Chr. 12: 54280690 - 54287088 on Build GRCh38
UniGene ID
Hs.546261
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031157.3 NP_112420.1 8 1001
XM_005268826.1 XP_005268883.1 8 995
GenBank mRNA BU541100.1 - 5 571
BX425482.2 - 8 925

View Full Product Details