Gene Transcripts
- Gene Symbol
- HSPA4
- Entrez Gene ID
- 3308
- Gene Name
- heat shock protein family A (Hsp70) member 4
- Gene Aliases
- APG-2, HEL-S-5a, HS24/P52, HSPH2, RY, hsp70, hsp70RY
- Chromosome Location
- Chr. 5: 133051970 - 133105017 on Build GRCh38
- UniGene ID
- Hs.90093
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002154.3 |
NP_002145.3
|
1, 2 |
381 |
|
GenBank mRNA |
AB023420.1 |
- |
1, 2 |
378 |
|
AB209151.1 |
- |
1, 2 |
302 |
|
AK294859.1 |
- |
1, 2 |
296 |
|
AK311097.1 |
- |
1, 2 |
112 |
|
BC002526.2 |
- |
1, 2 |
300 |
|
BC110861.1 |
- |
1, 2 |
241 |
6169048 |
BC126122.1 |
- |
1, 2 |
342 |
8991838 |
BC126124.1 |
- |
1, 2 |
342 |
8991840 |
BG723797.1 |
- |
1, 2 |
389 |
|
BT007375.1 |
- |
1, 2 |
100 |
|
FJ224293.1 |
- |
1, 2 |
378 |
|
L12723.1 |
- |
1, 2 |
233 |
|
View Full Product Details