Gene Transcripts
- Gene Symbol
- TCP11L1
- Entrez Gene ID
- 55346
- Gene Name
- t-complex 11 like 1
- Gene Aliases
- dJ85M6.3
- Chromosome Location
- Chr. 11: 33039417 - 33079454 on Build GRCh38
- UniGene ID
- Hs.655341
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145541.1 |
NP_001139013.1
|
9, 10 |
1546 |
|
NM_018393.3 |
NP_060863.3
|
9, 10 |
1730 |
|
XM_011520204.2 |
XP_011518506.1
|
9, 10 |
1561 |
|
XM_011520205.2 |
XP_011518507.1
|
9, 10 |
1729 |
|
XM_011520206.2 |
XP_011518508.1
|
9, 10 |
1747 |
|
XM_011520207.2 |
XP_011518509.1
|
9, 10 |
1743 |
|
XM_017017989.1 |
XP_016873478.1
|
9, 10 |
1744 |
|
XM_017017990.1 |
XP_016873479.1
|
9, 10 |
1714 |
|
XM_017017991.1 |
XP_016873480.1
|
7, 8 |
1107 |
|
XM_017017992.1 |
XP_016873481.1
|
7, 8 |
1165 |
|
GenBank mRNA |
AK002198.1 |
- |
9, 10 |
1528 |
|
AK021448.1 |
- |
4, 5 |
692 |
|
AK090646.1 |
- |
9, 10 |
1713 |
|
BC041696.1 |
- |
9, 10 |
1549 |
5756130 |
View Full Product Details