Gene Transcripts
- Gene Symbol
- NSUN5P1
- Entrez Gene ID
- 155400
- Gene Name
- NOP2/Sun RNA methyltransferase family member 5 pseudogene 1
- Gene Aliases
- NSUN5B, WBSCR20B
- Chromosome Location
- Chr. 7: 75410322 - 75416792 on Build GRCh38
- UniGene ID
- Hs.436034
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126486.1 |
- |
3 |
1729 |
|
View Full Product Details