Gene Transcripts
- Gene Symbol
- HNRNPA1
- Entrez Gene ID
- 3178
- Gene Name
- heterogeneous nuclear ribonucleoprotein A1
- Gene Aliases
- ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1
- Chromosome Location
- Chr. 12: 54280690 - 54287088 on Build GRCh38
- UniGene ID
- Hs.546261
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002136.3 |
NP_002127.1
|
1, 2 |
134 |
|
NM_031157.3 |
NP_112420.1
|
1, 2 |
134 |
|
NR_135167.1 |
|
1, 2 |
134 |
|
XM_005268826.1 |
XP_005268883.1
|
1, 2 |
128 |
|
XM_017019251.1 |
XP_016874740.1
|
1, 2 |
128 |
|
GenBank mRNA |
AK129814.1 |
- |
1, 2 |
76 |
|
AK291113.1 |
- |
1, 2 |
74 |
|
AK298176.1 |
- |
1, 2 |
77 |
|
AK303303.1 |
- |
1, 2 |
76 |
|
AK308679.1 |
- |
1, 2 |
76 |
|
BC002355.2 |
- |
1, 2 |
55 |
2821751 |
BC009600.1 |
- |
1, 2 |
77 |
4134638 |
BC012158.1 |
- |
1, 2 |
37 |
4564655 |
BC020442.2 |
- |
1 |
44 |
|
BC033714.1 |
- |
1, 2 |
66 |
4420039 |
BC052296.1 |
- |
1, 2 |
46 |
6297642 |
BC070315.1 |
- |
1, 2 |
76 |
4101267 |
BC071945.1 |
- |
1 |
66 |
5475712 |
BC073162.1 |
- |
1 |
60 |
6154972 |
BC074502.1 |
- |
1, 2 |
59 |
3455742 |
BC103707.1 |
- |
1, 2 |
44 |
6154800 |
BQ277208.1 |
- |
|
134 |
|
BX425482.2 |
- |
1, 2 |
58 |
|
CD705336.1 |
- |
1, 2 |
151 |
|
DC358304.1 |
- |
1, 2 |
230 |
|
U00947.1 |
- |
1, 2 |
24 |
|
X06747.1 |
- |
1, 2 |
101 |
|
X79536.1 |
- |
1, 2 |
42 |
|
- Gene Symbol
- HNRNPA1P10
- Entrez Gene ID
- 664709
- Gene Name
- heterogeneous nuclear ribonucleoprotein A1 pseudogene 10
- Gene Aliases
- HNRPA1L-2
- Chromosome Location
- Chr. 19: 11665736 - 11667089 on Build GRCh38
- UniGene ID
- Hs.534822
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002944.2 |
|
1 |
75 |
|
GenBank mRNA |
EC496431.1 |
- |
1 |
48 |
|
- Gene Symbol
- HNRNPA1P6
- Entrez Gene ID
- 729423
- Gene Name
- heterogeneous nuclear ribonucleoprotein A1 pseudogene 6
- Gene Aliases
- HNRNPA1L, HNRNPA1L1, HNRPA1L
- Chromosome Location
- Chr. 1: 58047849 - 58049373 on Build GRCh38
- UniGene ID
- Hs.657333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC121134.1 |
- |
1 |
60 |
|
- Gene Symbol
- SRR
- Entrez Gene ID
- 63826
- Gene Name
- serine racemase
- Gene Aliases
- ILV1, ISO1
- Chromosome Location
- Chr. 17: 2303371 - 2325267 on Build GRCh38
- UniGene ID
- Hs.461954
- Species
- Homo sapiens
- Gene Symbol
- XKRX
- Entrez Gene ID
- 402415
- Gene Name
- XK related, X-linked
- Gene Aliases
- XKR2, XPLAC, XRG2
- Chromosome Location
- Chr. X: 100886924 - 100957401 on Build GRCh38
- UniGene ID
- Hs.364911
- Species
- Homo sapiens
View Full Product Details