Gene Transcripts
- Gene Symbol
- ADAMTSL3
- Entrez Gene ID
- 57188
- Gene Name
- ADAMTS like 3
- Gene Aliases
- ADAMTSL-3
- Chromosome Location
- Chr. 15: 83654086 - 84039842 on Build GRCh38
- UniGene ID
- Hs.459162
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092324.1 |
- |
9 |
1863 |
|
- Gene Symbol
- KCNQ1OT1
- Entrez Gene ID
- 10984
- Gene Name
- KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
- Gene Aliases
- KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012
- Chromosome Location
- Chr. 11: 2608328 - 2699998 on Build GRCh38
- UniGene ID
- Hs.604823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002728.3 |
|
1 |
61372 |
|
- Gene Symbol
- KIAA1328
- Entrez Gene ID
- 57536
- Gene Name
- KIAA1328
- Gene Aliases
- Chromosome Location
- Chr. 18: 36828929 - 37232127 on Build GRCh38
- UniGene ID
- Hs.4045
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC008670.1 |
- |
2 |
972 |
|
View Full Product Details