Gene Transcripts
- Gene Symbol
- SMA4
- Entrez Gene ID
- 11039
- Gene Name
- glucuronidase beta pseudogene
- Gene Aliases
- SMA3, b55C20.2
- Chromosome Location
- Chr. 5: 70127462 - 70290177 on Build GRCh38
- UniGene ID
- Hs.582500
- Species
- Homo sapiens
- Gene Symbol
- GUSBP1
- Entrez Gene ID
- 728411
- Gene Name
- glucuronidase, beta pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 5: 21459480 - 21589372 on Build GRCh38
- UniGene ID
- Hs.88181
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027028.2 |
|
5 |
517 |
|
GenBank mRNA |
AK290693.1 |
- |
5 |
484 |
|
BC147000.1 |
- |
5 |
471 |
|
- Gene Symbol
- GUSBP2
- Entrez Gene ID
- 387036
- Gene Name
- glucuronidase, beta pseudogene 2
- Gene Aliases
- GUSBL1, GUSBP4, SMA3-L, SMAC3L, SMAC3L2, b55C20.1, bA239L20.1, bA239L20.5, bGLU-Lp
- Chromosome Location
- Chr. 6: 26871487 - 26956554 on Build GRCh38
- UniGene ID
- Hs.675815
- Species
- Homo sapiens
- Gene Symbol
- GUSBP3
- Entrez Gene ID
- 653188
- Gene Name
- glucuronidase, beta pseudogene 3
- Gene Aliases
- Chromosome Location
- Chr. 5: 69639463 - 69710527 on Build GRCh38
- UniGene ID
- Hs.631974
- Species
- Homo sapiens
- Gene Symbol
- GUSBP9
- Entrez Gene ID
- 100049076
- Gene Name
- glucuronidase, beta pseudogene 9
- Gene Aliases
- Chromosome Location
- Chr. 5: 71220226 - 71259295 on Build GRCh38
- UniGene ID
- Hs.665231
- Species
- Homo sapiens
- Gene Symbol
- SMA5
- Entrez Gene ID
- 11042
- Gene Name
- glucuronidase beta pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 5: 70481043 - 70585722 on Build GRCh38
- UniGene ID
- Hs.529793
- Species
- Homo sapiens
View Full Product Details