Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171192.1 | NP_001164663.1 | 2, 3 | 458 | |
NM_017711.3 | NP_060181.2 | 2, 3 | 458 | ||
GenBank mRNA | AB048363.1 | - | 2, 3 | 348 | |
AK000214.1 | - | 2, 3 | 170 | ||
AK301025.1 | - | 2, 3 | 282 | ||
AK307778.1 | - | 2, 3 | 159 | ||
AY358986.1 | - | 2, 3 | 153 | ||
BC032009.1 | - | 2, 3 | 155 | 4747934 |