Gene Transcripts

Gene Symbol
Nap1l2
Entrez Gene ID
317247
Gene Name
nucleosome assembly protein 1-like 2
Gene Aliases
Chromosome Location
Chr. X: 73997274 - 73999677 on Build Rnor_6.0
UniGene ID
Rn.43670
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024789.1 NP_001019960.1 1 2166
GenBank mRNA BC094525.1 - 1 2166

View Full Product Details