Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030792.6 | NP_110419.5 | 10 | 1277 | |
XM_006718697.3 | XP_006718760.1 | 11 | 1883 | ||
XM_011545276.2 | XP_011543578.1 | 12 | 1669 | ||
XM_011545277.2 | XP_011543579.1 | 12 | 2057 | ||
XM_011545278.2 | XP_011543580.1 | 12 | 2120 | ||
XM_011545279.2 | XP_011543581.1 | 10 | 1639 | ||
XM_011545280.1 | XP_011543582.1 | 9 | 972 | ||
XM_011545281.2 | XP_011543583.1 | 5 | 792 | ||
XM_011545285.2 | XP_011543587.1 | 6 | 2731 | ||
XM_011545286.1 | XP_011543588.1 | 5 | 528 | ||
XM_011545287.1 | XP_011543589.1 | 5 | 528 | ||
GenBank mRNA | AF318351.1 | - | 10 | 1133 | |
AF318377.1 | - | 10 | 1546 | ||
AF451987.1 | - | 3 | 294 | ||
AK074848.1 | - | 4 | 365 | ||
AK098808.1 | - | 5 | 528 | ||
AK310490.1 | - | 5 | 474 | ||
AL833941.1 | - | 11 | 1314 | ||
AY358477.1 | - | 5 | 544 | ||
BC018771.1 | - | 11 | 1325 | 4845520 | |
BC033391.1 | - | 6 | 2267 | 4830733 |