Gene Transcripts
- Gene Symbol
- ELF1
- Entrez Gene ID
- 1997
- Gene Name
- E74 like ETS transcription factor 1
- Gene Aliases
- Chromosome Location
- Chr. 13: 40931919 - 41061414 on Build GRCh38
- UniGene ID
- Hs.135646
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145353.1 |
NP_001138825.1
|
4, 5 |
685 |
|
NM_172373.3 |
NP_758961.1
|
5, 6 |
902 |
|
XM_005266276.1 |
XP_005266333.1
|
5, 6 |
791 |
|
XM_005266277.3 |
XP_005266334.1
|
5, 6 |
1334 |
|
XM_011534950.1 |
XP_011533252.1
|
7, 8 |
1140 |
|
XM_011534951.2 |
XP_011533253.1
|
6, 7 |
1453 |
|
XM_017020409.1 |
XP_016875898.1
|
7, 8 |
1120 |
|
XM_017020410.1 |
XP_016875899.1
|
6, 7 |
1057 |
|
XM_017020411.1 |
XP_016875900.1
|
6, 7 |
1003 |
|
XM_017020412.1 |
XP_016875901.1
|
6, 7 |
1131 |
|
XM_017020413.1 |
XP_016875902.1
|
6, 7 |
1185 |
|
GenBank mRNA |
AK304289.1 |
- |
4, 5 |
628 |
|
BC030507.2 |
- |
4, 5 |
757 |
4385989 |
M82882.1 |
- |
5, 6 |
844 |
|
View Full Product Details