Gene Transcripts

Gene Symbol
HNRNPA1
Entrez Gene ID
3178
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Gene Aliases
ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1
Chromosome Location
Chr. 12: 54280690 - 54287088 on Build GRCh38
UniGene ID
Hs.546261
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002136.3 NP_002127.1 10 1456
NM_031157.3 NP_112420.1 11 1612
XM_017019251.1 XP_016874740.1 9 1375
GenBank mRNA AK129814.1 - 10 1398
BC002355.2 - 10 1377 2821751
BC009600.1 - 10 1399 4134638
BC012158.1 - 10 1359 4564655
BC033714.1 - 10 1388 4420039
BC035253.1 - 6 815
BC052296.1 - 10 1368 6297642
BC074502.1 - 10 1381 3455742
BC103707.1 - 9 1207 6154800
U00947.1 - 10 1345
X06747.1 - 10 1422

Gene Symbol
XKRX
Entrez Gene ID
402415
Gene Name
XK related, X-linked
Gene Aliases
XKR2, XPLAC, XRG2
Chromosome Location
Chr. X: 100886924 - 100957401 on Build GRCh38
UniGene ID
Hs.364911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011530954.2 XP_011529256.1 4 3815

View Full Product Details