Gene Transcripts

Gene Symbol
Gckr
Entrez Gene ID
25658
Gene Name
glucokinase (hexokinase 4) regulator
Gene Aliases
GLRE
Chromosome Location
Chr. 6: 26355296 - 26385761 on Build Rnor_6.0
UniGene ID
Rn.7863
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013120.2 NP_037252.1 8 750
GenBank mRNA BC081843.1 - 8 750
KJ026952.1 - 8 573
X68497.1 - 8 594

View Full Product Details