Gene Transcripts
- Gene Symbol
- Anp32a
- Entrez Gene ID
- 11737
- Gene Name
- acidic (leucine-rich) nuclear phosphoprotein 32 family, member A
- Gene Aliases
- Anp32, I1PP2A, LANP, PHAP1, W91701, pp32
- Chromosome Location
- Chr. 9: 62341343 - 62378804 on Build GRCm38
- UniGene ID
- Mm.269088
- Species
- Mus musculus
- Gene Symbol
- Anp32-ps
- Entrez Gene ID
- 448829
- Gene Name
- acidic (leucine-rich) nuclear phosphoprotein 32 family, pseudogene
- Gene Aliases
- Anp32c, Anp32c-ps, Anp32cl, EG448829
- Chromosome Location
- Chr. 11: 33394905 - 33395397 on Build GRCm38
- UniGene ID
- Mm.159019
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY702092.1 |
- |
1 |
102 |
|
View Full Product Details