Gene Transcripts

Gene Symbol
2310057M21Rik
Entrez Gene ID
68277
Gene Name
RIKEN cDNA 2310057M21 gene
Gene Aliases
3110040E10Rik, AI429544
Chromosome Location
Chr. 7: 131342710 - 131362843 on Build GRCm38
UniGene ID
Mm.121122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026655.3 NP_080931.1 2 203
GenBank mRNA AK019097.2 - 2 205
AK088850.1 - 2 239
AK148008.1 - 212
BC044749.2 - 2 211 5102741
BC052432.1 - 2 201

View Full Product Details