Gene Transcripts
- Gene Symbol
- Atp11a
- Entrez Gene ID
- 50770
- Gene Name
- ATPase, class VI, type 11A
- Gene Aliases
- 4930558F19Rik, AU040868, Atpc1h, Ih
- Chromosome Location
- Chr. 8: 12757016 - 12868728 on Build GRCm38
- UniGene ID
- Mm.257837
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK028779.1 |
- |
1 |
1788 |
|
View Full Product Details