gRNA Design Details
- Target DNA Sequence
- TGCAAAGTAGCTGATAGAAC
- PAM Sequence
- GGG
- Target locus
-
Chr.1: 51477553 - 51477575 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
83.3/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Nabp1
- Entrez Gene ID
-
109019
- Gene Name
- nucleic acid binding protein 1
- Gene Aliases
-
4930434H03Rik, 4930442A21Rik, 4930488J04Rik, 4933440J18Rik, 5830411E10Rik, AI852561, Nbp1, Obfc2a, Ssb2
- Chromosome Location
-
Chr.1: 51465297 - 51478458 on GRCm38
- UniGene ID
- Mm.196290
- Species
-
Mouse