gRNA Design Details
- Target DNA Sequence
- TGGATATGCGCGAGAGCGAC
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 97142861 - 97142883 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.44/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Eogt
- Entrez Gene ID
-
101351
- Gene Name
- EGF domain-specific O-linked N-acetylglucosamine (GlcNAc) transferase
- Gene Aliases
-
A130022J15Rik, AI447490, AW214473, AW259391, Aer61
- Chromosome Location
-
Chr.6: 97110016 - 97149703 on GRCm38
- UniGene ID
- Mm.116802
- Species
-
Mouse