gRNA Design Details

Target DNA Sequence
GTCCACGTCTTCCATCTCCT
PAM Sequence
GGG
Target locus
Chr.1: 74284010 - 74284032 on GRCm38
Strand
Forward
Application
Gene Knockout
Score: 86.3/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Aamp
Entrez Gene ID
227290
Gene Name
angio-associated migratory protein
Gene Aliases
AU040907, Aamp-rs
Chromosome Location
Chr.1: 74279840 - 74284738 on GRCm38
UniGene ID
Mm.379371
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001190444.1 NP_001177373.1 2
NM_146110.3 NP_666222.2 2