gRNA Design Details
- Target DNA Sequence
- GTCTCTCCAAATAGTCTGAT
- PAM Sequence
- TGG
- Target locus
-
Chr.10: 13605661 - 13605683 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRPF18
- Entrez Gene ID
-
8559
- Gene Name
- pre-mRNA processing factor 18
- Gene Aliases
-
PRP18, hPrp18
- Chromosome Location
-
Chr.10: 13586811 - 13645279 on GRCh38
- UniGene ID
- Hs.660649
- Species
-
Human