Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005615.4 | NP_005606.1 | 2 | 716 | |
XM_017021566.1 | XP_016877055.1 | 3 | 603 | ||
XM_017021567.1 | XP_016877056.1 | 3 | 616 | ||
GenBank mRNA | AI151384.1 | - | 1 | 346 | |
AK313580.1 | - | 2 | 699 | ||
BC020848.1 | - | 2 | 716 | 4732021 | |
CR456965.1 | - | 1 | 423 |