Gene Transcripts
- Gene Symbol
- LOC100130587
- Entrez Gene ID
- 100130587
- Gene Name
- uncharacterized LOC100130587
- Gene Aliases
- Chromosome Location
- Chr. 20: 63359988 - 63371177 on Build GRCh38
- UniGene ID
- Hs.640129
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110634.1 |
|
4 |
1241 |
|
GenBank mRNA |
AK127527.1 |
- |
4 |
1241 |
|
View Full Product Details