Gene Transcripts

Gene Symbol
Aasdh
Entrez Gene ID
231326
Gene Name
aminoadipate-semialdehyde dehydrogenase
Gene Aliases
A230062G08Rik, A830035E16, Acsf4, U26
Chromosome Location
Chr. 5: 76873659 - 76905518 on Build GRCm38
UniGene ID
Mm.39271
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173765.3 NP_776126.1 13 2807
GenBank mRNA AB095954.1 - 13 2824
AK136849.1 - 13 2807
AK148405.1 - 12 2633
AK215263.1 - 2 102
BC058102.1 - 13 2792
BC094507.1 - 11 3123
BC128330.1 - 12 2679 40110405

View Full Product Details