Gene Transcripts

Gene Symbol
Olfr1294
Entrez Gene ID
258887
Gene Name
olfactory receptor 1294
Gene Aliases
MOR248-7
Chromosome Location
Chr. 2: 111537349 - 111540150 on Build GRCm38
UniGene ID
Mm.223297
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146885.1 NP_667096.1 1 742
GenBank mRNA BC127976.1 - 1 746

View Full Product Details