Gene Transcripts

Gene Symbol
Btnl2
Entrez Gene ID
547431
Gene Name
butyrophilin-like 2
Gene Aliases
BTL-II, BTLN2, Btl2, Gm315, NG9
Chromosome Location
Chr. 17: 34354789 - 34369492 on Build GRCm38
UniGene ID
Mm.441007
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_079835.2 NP_524574.1 9 1749
GenBank mRNA AK136521.1 - 9 1751
BC141103.1 - 9 1639 9055715

View Full Product Details