Gene Transcripts

Gene Symbol
Sp140
Entrez Gene ID
434484
Gene Name
Sp140 nuclear body protein
Gene Aliases
Chromosome Location
Chr. 1: 85600634 - 85645040 on Build GRCm38
UniGene ID
Mm.445821
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013817.2 NP_001013839.1 16 1941
GenBank mRNA BC069971.1 - 16 1941

View Full Product Details