Gene Transcripts
- Gene Symbol
- Gm2837
- Entrez Gene ID
- 100040552
- Gene Name
- spindlin family, member 2 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. X: 33056286 - 33057063 on Build GRCm38
- UniGene ID
- Mm.416917
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC117727.1 |
- |
1 |
148 |
|
- Gene Symbol
- Gm21637
- Entrez Gene ID
- 100862314
- Gene Name
- predicted gene, 21637
- Gene Aliases
- Chromosome Location
- Chr. X: 31960926 - 31962821 on Build GRCm38
- UniGene ID
- Mm.431329
- Species
- Mus musculus
- Gene Symbol
- Spin2-ps1
- Entrez Gene ID
- 74851
- Gene Name
- spindlin family, member 2, pseudogene 1
- Gene Aliases
- Gm14354, OTTMUSG00000016414
- Chromosome Location
- Chr. X: 3835879 - 3836656 on Build GRCm38
- UniGene ID
- Mm.431329
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029106.2 |
NP_083382.2
|
1 |
148 |
|
- Gene Symbol
- Spin2d
- Entrez Gene ID
- 100504429
- Gene Name
- spindlin family, member 2D
- Gene Aliases
- 4930408F14Rik
- Chromosome Location
- Chr. X: 73175302 - 73176989 on Build GRCm38
- UniGene ID
- Mm.307966
- Species
- Mus musculus
View Full Product Details