Gene Transcripts

Gene Symbol
Mepce
Entrez Gene ID
304361
Gene Name
methylphosphate capping enzyme
Gene Aliases
Chromosome Location
Chr. 12: 21721246 - 21725965 on Build Rnor_6.0
UniGene ID
Rn.198943
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100678.1 NP_001094148.1 1 1442

Gene Symbol
LOC100910540
Entrez Gene ID
100910540
Gene Name
7SK snRNA methylphosphate capping enzyme-like
Gene Aliases
Chromosome Location
Chr. 12: 21981675 - 21986474 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751154.3 XP_003751202.1 1 1522

View Full Product Details