Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014011.4 | NP_054730.1 | 2 | 1522 | |
NM_144949.2 | NP_659198.1 | 2 | 1610 | ||
GenBank mRNA | AB014571.1 | - | 2 | 1530 | |
AF073958.1 | - | 2 | 1610 | ||
AK290194.1 | - | 2 | 1536 | ||
AK296796.1 | - | 2 | 1480 | ||
AL136896.1 | - | 2 | 1522 | ||
BC032862.1 | - | 2 | 1623 | 5264754 |