Gene Transcripts
- Gene Symbol
- Prl3d2
- Entrez Gene ID
- 691552
- Gene Name
- Prolactin family 3, subfamily d, member 2
- Gene Aliases
- Pl-1
- Chromosome Location
- Chr. 17: 42849469 - 42855494 on Build Rnor_6.0
- UniGene ID
- Rn.198338
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ329282.1 |
- |
5 |
869 |
|
DQ329283.1 |
- |
5 |
869 |
|
View Full Product Details