Gene Transcripts
- Gene Symbol
- MAN2B2
- Entrez Gene ID
- 23324
- Gene Name
- mannosidase alpha class 2B member 2
- Gene Aliases
- Chromosome Location
- Chr. 4: 6575174 - 6622403 on Build GRCh38
- UniGene ID
- Hs.188464
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001292038.1 |
NP_001278967.1
|
1, 2 |
168 |
|
NM_015274.2 |
NP_056089.1
|
1, 2 |
168 |
|
GenBank mRNA |
AB023152.2 |
- |
1, 2 |
127 |
|
AK301473.1 |
- |
1, 2 |
153 |
|
AL832232.1 |
- |
1, 2 |
140 |
|
AL833071.2 |
- |
1, 2 |
141 |
|
BC033307.1 |
- |
1, 2 |
137 |
4932398 |
BC094773.1 |
- |
1, 2 |
168 |
30531368 |
BE243063.1 |
- |
1, 2 |
168 |
|
BM146210.1 |
- |
1, 2 |
168 |
|
View Full Product Details