Gene Transcripts
- Gene Symbol
- RNF19A
- Entrez Gene ID
- 25897
- Gene Name
- ring finger protein 19A, RBR E3 ubiquitin protein ligase
- Gene Aliases
- RNF19
- Chromosome Location
- Chr. 8: 100257059 - 100336218 on Build GRCh38
- UniGene ID
- Hs.292882
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001280539.1 |
NP_001267468.1
|
6, 7 |
1481 |
|
NM_015435.4 |
NP_056250.3
|
6, 7 |
1512 |
|
NM_183419.3 |
NP_904355.1
|
5, 6 |
1588 |
|
XM_005250853.3 |
XP_005250910.1
|
5, 6 |
1550 |
|
XM_011516956.2 |
XP_011515258.1
|
5, 6 |
1573 |
|
XM_017013302.1 |
XP_016868791.1
|
6, 7 |
1673 |
|
XM_017013303.1 |
XP_016868792.1
|
6, 7 |
1583 |
|
XM_017013304.1 |
XP_016868793.1
|
5, 6 |
1530 |
|
XM_017013305.1 |
XP_016868794.1
|
3, 4 |
1068 |
|
GenBank mRNA |
AB029316.1 |
- |
6, 7 |
1512 |
|
BC093938.1 |
- |
4, 5 |
1284 |
7939783 |
BC093940.2 |
- |
4, 5 |
1284 |
7939785 |
View Full Product Details