Gene Transcripts

Gene Symbol
Brs3
Entrez Gene ID
12209
Gene Name
bombesin-like receptor 3
Gene Aliases
BRS-3
Chromosome Location
Chr. X: 57043074 - 57048758 on Build GRCm38
UniGene ID
Mm.10687
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009766.3 NP_033896.2 2 871
GenBank mRNA AK038616.1 - 2 872
BC106961.1 - 2 852 40039080
BC106962.1 - 2 852 40039081

View Full Product Details