Gene Transcripts

Gene Symbol
Mnx1
Entrez Gene ID
15285
Gene Name
motor neuron and pancreas homeobox 1
Gene Aliases
HB9, Hlxb9, MNR2
Chromosome Location
Chr. 5: 29473823 - 29478470 on Build GRCm38
UniGene ID
Mm.103760
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019944.2 NP_064328.2 1 689
GenBank mRNA AF153046.1 - 689
BC132283.1 - 1 602 40130560
BC144718.1 - 1 602

View Full Product Details