Gene Transcripts

Gene Symbol
LTF
Entrez Gene ID
4057
Gene Name
lactotransferrin
Gene Aliases
GIG12, HEL110, HLF2, LF
Chromosome Location
Chr. 3: 46436005 - 46485234 on Build GRCh38
UniGene ID
Hs.529517
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199149.1 NP_001186078.1 4 558
NM_001321121.1 NP_001308050.1 4 669
NM_001321122.1 NP_001308051.1 7 902
NM_002343.5 NP_002334.2 4 669
GenBank mRNA AF332168.1 - 4 376
AK093852.1 - 7 905
AK290859.1 - 4 414
AK292813.1 - 4 414
AK298035.1 - 4 558
AK316330.1 - 4 478
AY137470.1 - 4 376
AY165046.1 - 4 388
AY178998.2 - 4 376
AY360320.1 - 4 377
AY493417.1 - 4 486
AY875691.1 - 4 376
BC015822.1 - 4 418 4251222
BC015823.1 - 4 420 4294752
BC022347.1 - 4 414 4251058
CD722125.1 - 4 402
DQ522304.1 - 4 376
EU779935.1 - 4 376
EU794669.1 - 4 411
KF992208.1 - 4 388
KT006756.1 - 4 376
M83202.1 - 4 383
M83205.1 - 4 362
M93150.1 - 4 393
U07643.1 - 4 473
X52941.1 - 4 370
X53961.1 - 4 670

View Full Product Details