Gene Transcripts

Gene Symbol
MEP1A
Entrez Gene ID
4224
Gene Name
meprin A subunit alpha
Gene Aliases
PPHA
Chromosome Location
Chr. 6: 46793357 - 46845987 on Build GRCh38
UniGene ID
Hs.179704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK225212.1 - 12 1687
AK315246.1 - 12 1669
BC131714.1 - 12 1668 40122874
M82962.1 - 12 1664

View Full Product Details