Gene Transcripts

Gene Symbol
ST3GAL4
Entrez Gene ID
6484
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Gene Aliases
CGS23, NANTA3, SAT3, SIAT4, SIAT4C, ST3GalIV, STZ
Chromosome Location
Chr. 11: 126355645 - 126459601 on Build GRCh38
UniGene ID
Hs.591947
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001254757.1 NP_001241686.1 5 485
NM_001254758.1 NP_001241687.1 5 358
NM_001254759.1 NP_001241688.1 3 502
NM_006278.2 NP_006269.1 5 684
XM_011542959.1 XP_011541261.1 4 287
XM_011542960.2 XP_011541262.1 4 287
XM_017018134.1 XP_016873623.1 5 370
GenBank mRNA AF035249.1 - 1 54
AF516602.1 - 3 212
AF516603.1 - 3 182
AF516604.1 - 3 212
AF525084.1 - 3 242
AK021929.1 - 3 186
AK291577.1 - 4 314
AK314738.1 - 4 284
AL520639.3 - 5 443
AL529887.3 - 5 359
AY040826.1 - 4 245
BC010645.1 - 5 358 3449625
BI259855.1 - 5 453
CR456858.1 - 4 215
DA210431.1 - 5 485
DA841563.1 - 3 503
L23767.1 - 3 224
X74570.1 - 5 377

View Full Product Details