Gene Transcripts

Gene Symbol
UBE2G1
Entrez Gene ID
7326
Gene Name
ubiquitin conjugating enzyme E2 G1
Gene Aliases
E217K, UBC7, UBE2G
Chromosome Location
Chr. 17: 4269217 - 4366674 on Build GRCh38
UniGene ID
Hs.741319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003342.4 NP_003333.1 5 893
GenBank mRNA BC002775.2 - 5 792
BC026288.1 - 5 701
BU161770.1 - 3 557
CD250951.1 - 3 593
D78514.1 - 5 553

View Full Product Details