Gene Transcripts
- Gene Symbol
- LOC105370109
- Entrez Gene ID
- 105370109
- Gene Name
- uncharacterized LOC105370109
- Gene Aliases
- Chromosome Location
- Chr. 13: 22883682 - 22909059 on Build GRCh38
- UniGene ID
- Hs.380803
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130366.1 |
- |
4 |
2331 |
|
- Gene Symbol
- NUS1
- Entrez Gene ID
- 116150
- Gene Name
- NUS1 dehydrodolichyl diphosphate synthase subunit
- Gene Aliases
- C6orf68, MGC:7199, NgBR, TANGO14
- Chromosome Location
- Chr. 6: 117675454 - 117710727 on Build GRCh38
- UniGene ID
- Hs.289008
- Species
- Homo sapiens
View Full Product Details