Gene Transcripts
- Gene Symbol
- LINC01341
- Entrez Gene ID
- 149134
- Gene Name
- long intergenic non-protein coding RNA 1341
- Gene Aliases
- Chromosome Location
- Chr. 1: 246789617 - 246791486 on Build GRCh38
- UniGene ID
- Hs.720494
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015422.1 |
|
2 |
650 |
|
GenBank mRNA |
AK022825.1 |
- |
5 |
2090 |
|
AK125177.1 |
- |
2 |
1175 |
|
View Full Product Details