Gene Transcripts
- Gene Symbol
- 4933405E24Rik
- Entrez Gene ID
- 71056
- Gene Name
- RIKEN cDNA 4933405E24 gene
- Gene Aliases
- Chromosome Location
- Chr. 11: 54177772 - 54213288 on Build GRCm38
- UniGene ID
- Mm.56332
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045506.1 |
|
1 |
300 |
|
GenBank mRNA |
AK016664.1 |
- |
1 |
303 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY716353.1 |
- |
1 |
303 |
|
Chr. 11 - 54177938 - 54213290 [-] |
View Full Product Details