Gene Transcripts

Gene Symbol
Pdap1
Entrez Gene ID
64527
Gene Name
PDGFA associated protein 1
Gene Aliases
Haspp28, PAP
Chromosome Location
Chr. 12: 11240775 - 11251002 on Build Rnor_6.0
UniGene ID
Rn.6158
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022595.1 NP_072117.1 5, 6 523
GenBank mRNA BC088140.1 - 5, 6 537
U26541.1 - 5, 6 523
U41744.1 - 5, 6 544

View Full Product Details