Gene Transcripts

Gene Symbol
Parp2
Entrez Gene ID
290027
Gene Name
poly (ADP-ribose) polymerase 2
Gene Aliases
Adprtl2
Chromosome Location
Chr. 15: 27739416 - 27749650 on Build Rnor_6.0
UniGene ID
Rn.22730
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106030.1 NP_001099500.1 16 1598
XM_006251856.2 XP_006251918.1 14 1442

View Full Product Details