Gene Transcripts

Gene Symbol
Ebf3
Entrez Gene ID
361668
Gene Name
early B-cell factor 3
Gene Aliases
Chromosome Location
Chr. 1: 209523153 - 209641198 on Build Rnor_6.0
UniGene ID
Rn.64435
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108506.1 NP_001101976.1 4 713
XM_006230462.2 XP_006230524.1 3 866
XM_006230463.2 XP_006230525.1 3 865
XM_006230464.2 XP_006230526.1 3 864
XM_006230465.2 XP_006230527.1 3 863
XM_008759997.1 XP_008758219.1 3 867
XM_008759998.1 XP_008758220.1 3 866
XM_008759999.1 XP_008758221.1 3 866
XM_008760000.1 XP_008758222.1 3 866
XM_008760001.1 XP_008758223.1 3 865
XM_008760002.1 XP_008758224.1 3 863

View Full Product Details