Gene Transcripts

Gene Symbol
Tmem9
Entrez Gene ID
66241
Gene Name
transmembrane protein 9
Gene Aliases
1500015G18Rik, AW545782
Chromosome Location
Chr. 1: 136008219 - 136035030 on Build GRCm38
UniGene ID
Mm.41773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160145.1 NP_001153617.1 2, 3 307
NM_001160146.1 NP_001153618.1 2, 3 203
NM_025439.3 NP_079715.1 2, 3 172
GenBank mRNA AK005248.1 - 2, 3 159
AK021225.1 - 2, 3 159
AK134909.1 - 2, 3 137
AK154423.1 - 2, 3 159
BC016524.1 - 2, 3 147 3594475
BC020291.1 - 1, 2 124
BC048017.1 - 2, 3 126
BQ918002.1 - 2, 3 394
CA460035.1 - 2, 3 209
CX208138.1 - 2, 3 203

View Full Product Details