Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000593.5 | NP_000584.2 | 11 | 2896 | |
NM_001292022.1 | NP_001278951.1 | 11 | 2156 | ||
GenBank mRNA | AK093980.1 | - | 1 | 2158 | |
AK123494.1 | - | 5 | 2483 | ||
AK130176.1 | - | 6 | 1308 | ||
AK302340.1 | - | 11 | 2127 | ||
BC014081.2 | - | 11 | 2896 | 4304716 | |
BX648013.1 | - | 11 | 2904 | ||
X57522.1 | - | 11 | 2769 |