Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271807.1 | NP_001258736.1 | 2 | 293 | |
NM_178174.3 | NP_835468.1 | 2 | 293 | ||
XM_017010822.1 | XP_016866311.1 | 4 | 483 | ||
XM_017010823.1 | XP_016866312.1 | 3 | 371 | ||
XM_017010824.1 | XP_016866313.1 | 3 | 377 | ||
XM_017010825.1 | XP_016866314.1 | 4 | 483 | ||
GenBank mRNA | AF508193.1 | - | 2 | 232 | |
AF534822.1 | - | 2 | 276 | ||
AF534823.1 | - | 2 | 276 | ||
AY358357.1 | - | 2 | 281 | ||
BC100944.1 | - | 2 | 276 | 40004190 | |
BC100945.1 | - | 2 | 276 | 40004191 |